missing translation for 'onlineSavingsMsg'
Learn More

Inhibin alpha Antibody (PO12/8), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP2-61936AF750

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213461

  • 551.82€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Inhibin alpha Monoclonal specifically detects Inhibin alpha in Human samples. It is validated for ELISA, Immunohistochemistry.
Especificaciones

Especificaciones

Inhibin alpha
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
INHA
Synthetic peptide corresponding to epitope region aa73-96 of the alphaC region subunit of Inhibin alpha
Primary
Human
IgG2a
ELISA, Immunohistochemistry
PO12/8
ELISA, Immunohistochemistry
A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha
Mouse
0.1 mL
3623
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales