Learn More
Influenza A Hemagglutinin H1 Antibody (InA97), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Bio-Techne NB100-73158R
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18215473
  • 402.80€ / 0.20 ml
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
Influenza A Hemagglutinin H1 Monoclonal antibody specifically detects Influenza A Hemagglutinin H1 in Virus samples. It is validated for Western Blot, ELISA.
Especificaciones
Especificaciones
Influenza A Hemagglutinin H1
Monoclonal
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
Protein A purified
RUO
Detects H1 of Influenza A as well as recombinant H1 with high specificity and does not interact with haemagglutinin H3.
Store at 4C in the dark.
IgG1
Western Blot, ELISA
InA97
Western Blot, ELISA, Sandwich ELISA
HA, Hemagglutinin, Hemagglutinin HA
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza A/NewCaledonia/20/99 (strain H1N1).
0.2 mL
Primary
Virus
Purified