missing translation for 'onlineSavingsMsg'
Learn More

Indole-4-carboxaldehyde, 97%, Thermo Scientific™

Product Code. 11307569
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11307569 5 g 5g
15413817 250 mg 250mg
11397559 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11307569 Supplier Thermo Scientific Alfa Aesar Supplier No. H28019.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Indolecarbaldehyde is a synthetic intermediate used for pharmaceutical synthesis. Also used as reactant in Biginelli reaction, synthesis of aurora kinase A inhibitors, preparation of antitumor agents, intermolecular Friedel-Crafts acylation, preparation of inhibitors of cell division in E. coli and synthesis of Hantzsch pyridine-containing Schiff bases.

Solubility
Soluble in ethanol and acetone. Insoluble in water.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1074-86-8
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD01632221
InChI Key JFDDFGLNZWNJTK-UHFFFAOYSA-N
Synonym indole-4-carboxaldehyde, 4-formylindole, 1h-indole-4-carboxaldehyde, indole-4-carbaldehyde, 4-indole-carboxaldehyde, 4-indolecarbaldehyde, 4-formyl-indole, 4-formyl-1h-indole, 1h-indol-4-carbaldehyde, indole-4-aldehyde
PubChem CID 333703
IUPAC Name 1H-indole-4-carbaldehyde
SMILES O=CC1=C2C=CNC2=CC=C1

Specifications

Melting Point 143°C to 144°C
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Soluble in ethanol and acetone. Insoluble in water.
Formula Weight 145.16
Percent Purity 97%
Chemical Name or Material Indole-4-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.