Learn More
Thermo Scientific Chemicals Indacaterol maleate
Product Code. 16524434 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16524434 1 g 1g
16534434 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16524434 Supplier Thermo Scientific Chemicals Supplier No. 464650010
Request Bulk or Custom Format
This item is not returnable. View return policy
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 753498-25-8
Molecular Formula C28H32N2O7
Molecular Weight (g/mol) 508.57
MDL Number MFCD20526769,MFCD18782702
InChI Key IREJFXIHXRZFER-UHFFFAOYNA-N
IUPAC Name 5-{2-[(5,6-diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl}-8-hydroxy-1,2-dihydroquinolin-2-one; but-2-enedioic acid
SMILES OC(=O)C=CC(O)=O.CCC1=C(CC)C=C2CC(CC2=C1)NCC(O)C1=C2C=CC(=O)NC2=C(O)C=C1
Specifications
Melting Point 195.0°C to 197.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in dmso
Formula Weight 508.57
Chemical Name or Material Indacaterol maleate