missing translation for 'onlineSavingsMsg'
Learn More

IMPA1 Antibody (2G5), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP2-59416AF647

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216040

  • 562.62€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

IMPA1 Monoclonal antibody specifically detects IMPA1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Especificaciones

Especificaciones

IMPA1
Monoclonal
Alexa Fluor 647
EC 3.1.3.25, IMP, IMP 1, IMPA, IMPase 1, inositol monophosphatase 1, inositol(myo)-1(or 4)-monophosphatase 1, Inositol-1(or 4)-monophosphatase 1, Lithium-sensitive myo-inositol monophosphatase A1
Recombinant human IMPA1 (1-277aa) purified from E. coli
0.1 mL
Lipid and Metabolism
3612
Store at 4°C in the dark.
IgG1 κ
Western Blot, Flow Cytometry, ELISA, Immunofluorescence
2G5
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales