Learn More
Hughes Immersion Heated 1500 L Tank Shower
Product Code. 17782348
Change view
Click to view available options
Capacity (Metric):
1500 L
Unit Size:
Each
Product Code. Capacity (Metric) Unit Size
17782348 1500 L Each
1 options
This item is not returnable. View return policy
Product Code. 17782348 Supplier Hughes Supplier No. HSHM14K1SS
  • ONLINE EXCLUSIVE PRICE
    21185.25€
    10005.00€ / paquete de 1
    Ahorro 11180.25€
    52% Descuento
 Restricted Item
Fecha estimada de envoi: 26-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Freeze protected emergency tank shower for use in non-hazardous environments. Designed to meet and exceed ANSI standards.
  • Immersion heated emergency tank shower suitable for use in non-hazardous environments
  • Designed to meet and exceed ANSI standards by providing a guaranteed flow of 76 liters per minute for 15 minutes, even when there is no mains water supply
  • Eye/face wash unit mounted internally
Specifications
Capacity (Metric) 1500 L
Material Stainless Steel
Product Type Tank Shower