Learn More
Iloprost-d4 (Major), TRC
Product Code. 30390820
Change view
Click to view available options
Quantity:
0.25 mg
Unit Size:
0.25mg
Product Code. Quantity Unit Size
30390820 0.25 mg 0.25mg
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30390820 Supplier Toronto Research Chemicals Supplier No. TRCI2673020.25MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Iloprost-d4
CAS 1035094-10-0
Molecular Formula C22H28D4O4
Quantity 0.25 mg
Synonym (5E)-5-[(3aS,4R,5R,6aS)-Hexahydro-5-hydroxy-4-[(1E,3S)-3-hydroxy-4-methyl-1-octen-6-yn-1-yl]-2(1H)-pentalenylidene]pentanoic Acid-d4, (1S,2R,3R,5S)-7-[(E)-4-Carboxybutylidene]-2-[(3S,1E)-3-hydroxy-4-methyl-6-octyn-1-enyl]-bicyclo[3.3.0]octane-d4, Ciloprost-d4, Endoprost-d4, Ilomedin-d4, ZK 36374-d4
InChI Formula InChI=1S/C22H32O4/c1-3-4-7-15(2)20(23)11-10-18-19-13-16(8-5-6-9-22(25)26)12-17(19)14-21(18)24/h8,10-11,15,17-21,23-24H,5-7,9,12-14H2,1-2H3,(H,25,26)/b11-10+,16-8+/t15?,17-,18+,19-,20+,21+/m0/s1/i5D2,6D2
SMILES O[C@@H](C(C)CC#CC)/C=C/[C@H]1[C@H](O)C[C@H]2[C@@H]1C/C(C2)=C/C([2H])([2H])C([2H])([2H])CC(O)=O
Molecular Weight (g/mol) 364.51