missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IL5 (Mouse) ELISA Kit

Marca:  Abnova™ KA3065.1Kit

Detalles adicionales : Peso : 0.02000kg

 Ver más versiones de este producto

Código de producto. 16057805

  • 578.00€ / 1 set


Fecha estimada de envío
17-02-2020
Añadir a la cesta

Includes: One 96-well plate pre-coated with anti-Mouse IL-5 antibody, 96 (8 x 12) wells
Lyophilized Mouse IL-5 standards, 10ng x 2
Sample/Standard diluent buffer, 30mL
Biotin conjugated anti-Mouse IL-5 antibody (Concentrated), Dilution: 1:100, 130μL
Antibody diluent buffer, 12mL
Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100, 130μL
ABC diluent buffer, 12mL
TMB substrate, 10mL
Stop solution, 10mL
Wash buffer (25X), 30mL

Descripción

Descripción

This kit is based on sandwich enzyme-linked immune-sorbent assay technology
Especificaciones

Especificaciones

15.6 to 1000pg/mL
Mouse
2pg/mL
Quantification
One 96-well plate pre-coated with anti-Mouse IL-5 antibody (96 (9 x 12) wells), Lyophilized Lyophilized Mouse IL-5 standards Mouse IL-5 standards (10ng x 2), Sample/Standard diluent buffer (30mL), Biotin conjugated anti-Mouse IL-5 antibody (Concentrated), Dilution: 1:100 (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 (130uL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X): 30mL
RUO
100μL
Store at 4°C for 4 months, or at -20°C for 8 months.
100μL
Biotin
Il5 (Mouse) ELISA Kit is intended for the quantitative measurement of mouse Il5.
Colorimetric
96 wells
Il5 (Mouse) ELISA Kit
Body Fluid, Cell Culture Supernate, Serum, Tissue Lysate
<2pg/mL
96 Assays
Videos
Documentos