Learn More
Abnova™ IL4 (Human) Recombinant Protein
Human IL4 (130 a.a.) full-length recombinant protein with His tag expressed in Barley grain (Hordeum vulgare).
Marca:  Abnova™ P5823.100ug
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 16202310
  • 549.00€ / 100 microgramos
Fecha estimada de envío
22-10-2020
Añadir a la cesta
Especificaciones
Especificaciones
NP_000580
Lyophilized
interleukin 4
SDS-PAGE Stained with Coomassie Blue
Store at -20°C on dry atmosphere. After reconstitution with sterile water to a concentration of no less than 100 ug/mL, store at -20°C. For long term storage it is recommended to add a carrier protein (0.1% HSA or BSA). Aliquot to avoid repeated freezing and thawing.
IL4
His
100 μg/mL
3565
Barley grain (Hordeum vulgare) expression system
100 μg
BCGF-1, BCGF1, BSF1, IL-4, MGC79402
Plants
Lyophilized from PBS, pH 7.2