missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL2 Mouse anti-Human, Clone: 2F11C5, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60306-1-IG-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.Especificaciones
IL2 | |
Monoclonal | |
2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P60568 | |
IL2 | |
IL2 Fusion Protein Ag10199 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
2F11C5 | |
Unconjugated | |
IL2 | |
IL 2, IL2, interleukin 2, lymphokine, T cell growth factor, TCGF | |
Mouse | |
Protein A | |
RUO | |
3558 | |
-20°C | |
Liquid |