missing translation for 'onlineSavingsMsg'
Learn More

IL17RC Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 15648-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16814914

  • 109.00€ / 20 microlitros

Envío estimado: 28-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a single-pass type I membrane protein that shares similarity with the interleukin-17 receptor (IL-17RA). Unlike IL-17RA, which is predomitly expressed in hemopoietic cells, and binds with high affinity to only IL-17A, this protein is expressed in nonhemopoietic tissues, and binds both IL-17A and IL-17F with similar affinities. The proinflammatory cytokines, IL-17A and IL-17F, have been implicated in the progression of inflammatory and autoimmune diseases. Multiple alternatively spliced transcript variants encoding different isoforms have been detected for this gene, and it has been proposed that soluble, secreted proteins lacking transmembrane and intracellular domains may function as extracellular antagonists to cytokine signaling.
Especificaciones

Especificaciones

IL17RC
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8K4C2, Q8NAC3
IL 17 receptor C, IL 17RC, IL 17RL, IL17 RL, IL17RC, IL17Rhom, IL17RL, interleukin 17 receptor C, ZcytoR14
Rabbit
IgG
20 μL
-20°C
Primary
171095, 84818
Western Blot
Unconjugated
IL17RC
Liquid
Il17rc
IL17RC Fusion Protein Ag8190
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human
Videos
SDS
Documentos
Certificados