missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IL17A (Human) ELISA Kit

In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Human IL-17A

Marca:  Abnova™ KA1732.1Kit

Detalles adicionales : Peso : 0.02000kg

 Ver más versiones de este producto

Código de producto. 16056765

  • 578.00€ / 1 set


Fecha estimada de envío
25-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

Biotin
IL17A (Human) ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IL17A.
Colorimetric
96-well Antibody coated microwell, 25mL Wash Buffer 20X concentrate, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B, 2 vials detection Antibody MSP, 8μL HRP-Streptavidian concentrate, 12mL TMB one-step Substrate reagent, 8m
96 Assays
Store the kit at -20°C. Standard should be stored at -20°C or -80°C. Avoid repeated freezing and thawing.
100μL
Human
10pg/mL
Quantification
IL17A (Human) ELISA Kit
Cell Culture Supernatant, Plasma, Serum, Urine
96 Assays
Videos
Documentos