missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-4 Rat, APC, Clone: MP4-25D2, enQuireBio™
Rat Monoclonal Antibody
Marca: EnquireBio QAB82-APC-25Tests
Detalles adicionales : Peso : 0.01000kg
Especificaciones
IL-4 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
IL4 | |
Protein A/G Purified | |
RUO | |
3565 | |
IgG1 κ |
Flow Cytometry | |
MP4-25D2 | |
APC | |
NP_758858 | |
Rat | |
25 Tests | |
Primary | |
Store at 2-4°C. |