missing translation for 'onlineSavingsMsg'
Learn More

IL-27 Mouse anti-Human, Clone: 1E1H5, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 66164-1-IG-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817795

  • 140.00€ / 20 microlitros
Fecha estimada de envío: 17-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by IL-27 is one of the subunits of a heterodimeric cytokine complex. This protein is related to interleukin 12A (IL12A). It interacts with Epstein-Barr virus induced gene 3 (EBI3), a protein similar to interleukin 12B (IL12B), and forms a complex that has been shown to drive rapid expansion of naive but not memory CD4(+) T cells. The complex is also found to synergize strongly with interleukin 12 to trigger interferon gamma (IFNG) production of naive CD4(+) T cells. The biological effects of this cytokine are mediated by the class I cytokine receptor (WSX1/TCRR).
Especificaciones

Especificaciones

IL-27
Monoclonal
1.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8NEV9
IL27
IL-27 Fusion Protein Ag21436
20 μL
Primary
Human
Antibody
IgG2b
Western Blot, Immunohistochemistry (Paraffin)
1E1H5
Unconjugated
IL27
IL 27, IL 27 A, IL 27 subunit alpha, IL 27A, IL27, IL27 A, IL27A, IL27p28, IL30, interleukin 27, Interleukin 27 subunit alpha
Mouse
Protein A
RUO
246778
-20°C
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales