missing translation for 'onlineSavingsMsg'
Learn More

IL-15R alpha Antibody (RM0103-6C38), Biotin, Novus Biologicals™

Rat Monoclonal Antibody

Marca:  Novus Biologicals NBP1-22466B

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213595

  • 590.70€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

IL-15R alpha Monoclonal specifically detects IL-15R alpha in Mouse samples. It is validated for Western Blot.
Especificaciones

Especificaciones

IL-15R alpha
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
IL15RA
Mouse recombinant IL-15R alpha (extracellular domain)
0.1 mL
3601
Mouse
Purified
Western Blot
RM0103-6C38
Western Blot
CD215 antigen, IL-15 receptor subunit alpha, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179
Rat
Protein A or G purified
Primary
This was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein of the extracellular domain of IL-15RA.
Store at 4C in the dark.
IgG2
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales