missing translation for 'onlineSavingsMsg'
Learn More

IL-12 R beta 1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP1-32449

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215970

  • 535.62€ / 100 microlitros
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

IL-12 R beta 1 Polyclonal specifically detects IL-12 R beta 1 in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

IL-12 R beta 1
Polyclonal
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
IL12RB1
Recombinant protein encompassing a sequence within the center region of human IL12 Receptor beta1. The exact sequence is proprietary.
Affinity Purified
RUO
Primary
Tested in human. Not yet tested in any other species.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot
1.0mg/mL
Western Blot 1:500-1:3000
CD212, CD212 antigen, IL-12 receptor beta component, IL-12 receptor subunit beta-1, IL12R, IL-12R subunit beta-1, IL12RB, IL-12RB1, IL-12R-beta-1, IL-12R-BETA1, interleukin 12 receptor, beta 1, interleukin-12 receptor beta-1 chain, interleukin-12 receptor subunit beta-1, MGC34454
Rabbit
73 kDa
100 μL
Cell Cycle and Replication
3594
Human
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only