Learn More
Invitrogen™ IL-10RA (CD210a) Human ELISA Kit
Sandwich ELISA Kit
652.00€ - 4715.00€
Especificaciones
Número de acceso | Q13651 |
---|---|
Sensibilidad del ensayo | 2,5 ng |
ensayo | ELISA sandwich de fase sólida |
Conjugado | Biotina |
Tipo de producto | Kit de ELISA |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15465023
|
Invitrogen™
EHIL10RA |
96 pruebas |
652.00€
1 set |
Fecha estimada de envío: 17-05-2024
Inicie sesión para ver el stock |
|||||
17814210
|
Invitrogen™
EHIL10RAX10 |
10 × 96 pruebas |
4715.00€
paquete de 10 |
Fecha estimada de envío: 27-05-2024
Inicie sesión para ver el stock |
|||||
17195469
|
Invitrogen™
EHIL10RAX5 |
5 × 96 pruebas |
2635.00€
cada uno |
Fecha estimada de envío: 24-05-2024
Inicie sesión para ver el stock |
|||||
Description
The Human Interleukin 10 Receptor Alpha (CD210a) (Hu IL-10RA) ELISA quantitates Hu IL-10RA in human serum, plasma or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-10RA. Principle of the method The Human IL-10RA solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The protein encoded by this gene is a receptor for interleukin 10. This protein is structurally related to interferon receptors. It has been shown to mediate the immunosuppressive signal of interleukin 10, and thus inhibits the synthesis of proinflammatory cytokines. This receptor is reported to promote survival of progenitor myeloid cells through the insulin receptor substrate-2/PI 3-kinase/AKT pathway. Activation of this receptor leads to tyrosine phosphorylation of JAK1 and TYK2 kinases. Two transcript variants, one protein-coding and the other not protein-coding, have been found for this gene.Specifications
Q13651 | |
ELISA sandwich de fase sólida | |
Kit de ELISA | |
Plasma, Sobrenadante, Suero | |
Humano | |
Lector de microplacas colorimétrico | |
< 12 % | |
Peroxidasa de rábano picante (HRP) | |
Transducción de señal | |
Humano | |
4 h 45 min |
2,5 ng | |
Biotina | |
Otras proteínas | |
Este kit de ELISA no muestra reactividad cruzada con las siguientes citoquinas probadas: angiogenina humana, BDNF, BLC, CNTF, ENA-78, FGF-4, IL-1 alfa, IL-1 beta, IL-2, IL-4, IL-5, IL-6, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, FGF-4, FGF-6, FGF-7, G-CSF, GDNF, GM-CSF, IFN-gamma, IGFBP-2, IGFBP-3, IGFBP-4, leptina (OB), MCP-1, MCP-2, MCP-3, MDC, MIF, MIG, MIP-1 alfa, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF,SDF-1 alfa, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alfa, TNF-beta, TPO, VEGF. | |
ELISA, ensayos de proteínas, biología de proteínas, biología del cáncer, inmunología, neurociencia, neurología, detección de proteínas, análisis de proteínas | |
3587 | |
< 10 % | |
RUO | |
De 2 °C a 8 °C | |
1 h 20 min |