missing translation for 'onlineSavingsMsg'
Learn More

IKBKG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 18474-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16818504

  • 109.00€ / 20 microlitros

Envío estimado: 28-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

IKK gamma is the regulatory subunit of the inhibitor of the kappaB kinase (IKK) complex, which activates NF-kappaB resulting in activation of genes involved in inflammation, immunity, cell survival, and other pathways. Mutations in this gene result in incontinentia pigmenti, hypohidrotic ectodermal dysplasia, and several other types of immunodeficiencies. Multiple transcript variants encoding different isoforms have been found for this gene. A pseudogene highly similar to this locus is located in an adjacent region of the X chromosome.
Especificaciones

Especificaciones

IKBKG
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O88522, Q6TMG5, Q9Y6K9
AMCBX1, FIP 3, FIP3, Fip3p, I kappa B kinase subunit gamma, IkB kinase subunit gamma, IKBKG, IKK gamma, IKKAP1, IKKG, IP, IP1, IP2, IPD2, NEMO, NF kappa B essential modifier, NF kappa B essential modulator
Rabbit
IgG
20 μL
-20°C
Primary
16151, 309295, 8517
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
IKBKG
Liquid
Ikbkg
IKBKG Fusion Protein Ag13358
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados