Learn More
Stainless Steel Strap Clamp for IKA Eurostar Power Control-Visc P4 and P7 Stirrers
Product Code. 10618972 Shop All IKA Products
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
Product Code. Quantity Unit Size
10618972 1 Each Each
1 options
Product Code. 10618972 Supplier IKA Supplier No. 0003159000
  • ONLINE EXCLUSIVE PRICE
    540.00€
    446.00€ / paquete de 1
    Ahorro 94.00€
    17% Descuento
En stock
Add to basket
Edge
Add to basket
Use this stainless steel clamp for securing vessels against walls or synchronizing rotation during stirring. IKA™ Stainless Steel Strap Clamps are designed for IKA Eurostar Power Control-Visc P4 and P7 Stirrers
  • Includes cross collar joint for Boss Head 14-259-123 and fastening angle bar
  • For use with stands 14-259-86 and 14-259-039
  • Suitable for vessel from 1 3/8 to 11 in. (40 to 330mm) dia.
TRUSTED_SUSTAINABILITY
Specifications
Includes Boss Head Clamp R 270
Quantity 1 Each
Type RH 5 Strap Clamp