missing translation for 'onlineSavingsMsg'
Learn More

IgM Monoclonal Antibody (II/41), Super Bright™ 436, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

en la oferta

Especificaciones

Antígeno IgM
Clon II/41
Concentración 0.2 mg/mL
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Ver más especificaciones

Descripción

Descripción

Rat Monoclonal Antibody

IgM Monoclonal antibody specifically detects IgM in Mouse samples. It is validated for Flow Cytometry.
Especificaciones

Especificaciones

IgM
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with BSA and 0.09% sodium azide; pH 7.2
16019
Primary
4° C, store in dark, DO NOT FREEZE!
II/41
Flow Cytometry
Super Bright 436
Rat
Mouse
P01872
IgG2a κ
Affinity chromatography
Antibody
Videos
SDS
Documentos

Documentos

Product Certifications