missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG1 Rat, Unconjugated, Clone: RTK2071, Isotype Control, STEMCELL Technologies
Rat Monoclonal Antibody
58.05€ - 194.40€
Specifications
Antígeno | IgG1 |
---|---|
Clon | RTK2071 |
Aplicaciones | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
Clasificación | Monoclonal |
Conjugado | Unconjugated |
Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
17108891
|
Stemcell Technologies
60075.1 |
50 μg |
58.05€
50 microgramos |
Fecha estimada de envío: 13-05-2024
Inicie sesión para ver el stock |
|||||
17198881
|
Stemcell Technologies
60075 |
500 μg |
194.40€
500 microgramos |
Fecha estimada de envío: 24-04-2024
Inicie sesión para ver el stock |
|||||
Description
- The RTK2071 antibody (IgG1, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody
- Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used
- If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen
- The use of an appropriate isotype control helps confirm the specificity of the antigen-specific antibody and indicates non-specific binding that may result from binding to Fc receptors or other cell components
- This antibody clone has been verified for use as an isotype control antibody for assessing non-specific binding to cells in applications such as flow cytometry and immunofluorescence microscopy applications (surface and intracellular staining)
Specifications
IgG1 | |
Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
IgG1 κ |
RTK2071 | |
Monoclonal | |
Rat | |
Isotype Control (Primary) |