Learn More
IgG1 Antibody (BIG715A), Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Bio-Techne NBP2-61074
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18213942
  • 308.85€ / 0.10 miligramo
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
IgG1 Monoclonal specifically detects IgG1 in Bovine, Goat, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Especificaciones
Especificaciones
IgG1
Monoclonal
1 mg/mL
Flow Cytometry 1:10 - 1:1000
IgG1, Igh-4, RGD1359539, VH7183
Bovine immunoglobulins
0.1 mg
Bovine, Goat, Sheep
IgG1
Flow Cytometry, Immunohistochemistry, Functional Assay
BIG715A
Unconjugated
Ascites with 0.09% Sodium Azide
Mouse
Unpurified
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.