Learn More
Igepal CO-520, TRC
Product Code. 30394775
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30394775 50 g 50g
30399345 10 g 10g
30389903 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394775 Supplier Toronto Research Chemicals Supplier No. TRCI26505050G
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 68412-54-4
Molecular Formula (C2H4O)n.C15H24O
Synonym ?-(Nonylphenyl)-?-hydroxy-poly(oxy-1,2-ethanediyl) Branched Polymer, Manual Registration, Generic Registration
Specifications
Quantity 50 g
Chemical Name or Material 4-Nonylphenol-ethoxylate (penta-) (technical) (branched)