missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ IgE (ε -Heavy Chain), Rabbit, Polyclonal, Epredia™

IgE (ε-Heavy Chain), Rb PAb is one marker in a comprehensive ab portfolio that addresses all major disease states, ensuring the ideal tests are always available.

Marca:  Epredia™ RB-1435-A1

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12682897

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Research Areas: Immunoglobulins; The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Host Species: Rabbit

Clone: Polyclonal

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: IgE isolated from a pool of normal human sera

Molecular Weight: 50kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Especificaciones

Especificaciones

IgE (epsilon-Heavy Chain)
Polyclonal
Purified Ab with BSA and Azide
IgE isolated from a pool of normal human sera
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
500 μL
Immunoglobulins
Human
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales