missing translation for 'onlineSavingsMsg'
Learn More

IgE Antibody (4H10), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NB500-472APCCY7

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214315

  • 402.80€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

IgE Monoclonal specifically detects IgE in Human samples. It is validated for Flow Cytometry.
Especificaciones

Especificaciones

IgE
Monoclonal
APC-Cyanine7
PBS with 0.05% Sodium Azide
IGHE
Purified human IgE.
0.1 mL
Primary
The antibody 4H10 reacts with human IgE; it recognizes an epitope different from the ones recognized by BE5 and 4G7 antibodies to IgE.
Store at 4°C in the dark. Do not freeze.
Flow Cytometry
4H10
Flow Cytometry
IgE, immunoglobulin heavy constant epsilon
Mouse
Ion exchange chromatography
Adaptive Immunity, Immunology
3497.0
Human
IgG1
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales