missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IgD (Human) ELISA Kit

highly sensitive two-site enzyme-linked immunoassay for the quantitative determination of IgD in Human biological samples.

Marca:  Abnova™ KA3824.1Kit

Detalles adicionales : Peso : 0.02000kg

 Ver más versiones de este producto

Código de producto. 16058255

  • 418.00€ / 1 set


Fecha estimada de envío
19-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

Human
Colorimetric
IgD (Human) ELISA Kit
1 Kit
50μL
IgD (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IgD.
Quantification
IgD (Human) ELISA kit
Cell Culture Supernatant, Milk, Plasma, Saliva, Serum, Urine
Store the kit at 4°C. For long term storage, please store at -20°C.
Videos
Documentos