missing translation for 'onlineSavingsMsg'
Learn More

IFN gamma Rat anti-Mouse, Alexa Fluor® 700, Clone: XMG1.2, eBioscience™

Rat Monoclonal Antibody

Marca:  Invitrogen 56-7311-82

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923324

  • 217.00€ / 100 microgramos


Fecha estimada de envío
25-08-2020
Añadir a la cesta

Descripción

Descripción

Excitation: 633-647 nm; Emission: 723 nm; Laser: Red Laser IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.
Especificaciones

Especificaciones

IFN gamma
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P01580
Ifng
IgG1, kappa
100 μg
4° C, store in dark, DO NOT FREEZE!
Primary
15978
Flow Cytometry
XMG1.2
Alexa Fluor™ 700
Ifng
Liquid
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Videos
Documentos