missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ IER2 Recombinant Protein

335.00€ - 508.00€

Specifications

Número de acceso AAH03625
Para utilizar con (aplicación) Antibody Production, Protein Array, ELISA, Western Blot
Formulación 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
ID de gen (Entrez) 9592
Peso molecular 50.6
View More Specs

Products 2
Product Code Brand Cantidad Price Quantity & Availability  
Product Code Brand Cantidad Price Quantity & Availability  
16103902
missing translation for 'viewDocuments' Promotion Details
Abnova™
H00009592-P01.10UG
10 ug
335.00€
10 microgramos
View alternative products
missing translation for 'toSaveUpto' 
»
Fecha estimada de envío: 30-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16113902
missing translation for 'viewDocuments' Promotion Details
Abnova™
H00009592-P01.25UG
25 ug
508.00€
25 microgramos
View alternative products
missing translation for 'toSaveUpto' 
»
Fecha estimada de envío: 30-05-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH03625
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
50.6
8
Glutathione Sepharose 4 Fast Flow
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IER2
Human
Recombinant
Solution
Antibody Production, Protein Array, ELISA, Western Blot
9592
IER2 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
MEVQKEAQRIMTLSVWKMYHSRMQRGGLRLHRSLQLSLVMRSARELYLSAKVEALEPEVSLPAALPSDPRLHPPREAESTAETATPDGEHPFPEPMDTQEAPTAEETSACCAPRPAKVSRKRRSSSLSDGGDAGLVPSKKARLEEKEEEEGASSEVADRLQPPPAQAEGAFPNLARVLQRRFSGLLNCSPAAPPTAPPACEAKPACRPADSMLNVLVRAVVAF
ETR101/MGC111472/MGC15265
IER2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates