missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ IDH3G (Human) Recombinant Protein
Human IDH3G full-length ORF ( AAH00933, 1 a.a. - 393 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00003421-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MALKVATVAGSAAKAVLGPALLCRPWEVLGAHEVPSRNIFSEQTIPPSAKYGGRHTVTMIPGDGIGPELMLHVKSVFRHACVPVDFEEVHVSSNADEEDIRNAIMAIRRNRVALKGNIETNHNLPPSHKSRNNILRTSLDLYANVIHCKSLPGVVTRHKDIDILIVRENTEGEYSSLEHESVAGVVESLKIITKAKSLRIAEYAFKLAQESGRKKVTAVHKANIMKLGDGLFLQCCREVAARYPQITFENMIVDNTTMQLVSRPQQFDVMVMPNLYGNIVNNVCAGLVGGPGLVAGANYGHVYAVFETATRNTGKSIANKNIANPTATLLASCMMLDHLKLHSYAASIRKAVLASMDNENMHTPDIGGQGTTSEAIQDVIRHIRVINGRAVEA
Especificaciones
AAH00933 | |
isocitrate dehydrogenase 3 (NAD+) gamma | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IDH3G | |
GST |
3421 | |
Wheat germ expression system | |
10 μg | |
H-IDHG | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |