missing translation for 'onlineSavingsMsg'
Learn More

IDH1 Mouse anti-Human, Clone: 2A6A2, Proteintech

Mouse Monoclonal Antibody

140.00€ - 399.00€

Especificaciones

Antígeno IDH1
Clon 2A6A2
Concentración 1 mg/mL
Aplicaciones Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Clasificación Monoclonal
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16827855
Ver documentos Detalles de la promoción
Proteintech
66197-1-IG-20UL
20 μL
140.00€
20 microlitros
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 17-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16817855
Ver documentos Detalles de la promoción
Proteintech
66197-1-IG-150UL
150 μL
399.00€
150 microlitros
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 17-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This nuclear enzyme is involved in processes such as chromosome condensation, chromatid separation, and the relief of torsional stress that occurs during DNA transcription and replication. It catalyzes the transient breaking and rejoining of two strands of duplex DNA which allows the strands to pass through one another, thus altering the topology of DNA. Two forms of this enzyme exist as likely products of a gene duplication event. The gene encoding this form, beta, is localized to chromosome 3 and the alpha form is localized to chromosome 17. The gene encoding this enzyme functions as the target for several anticancer agents and a variety of mutations in this gene have been associated with the development of drug resistance. Reduced activity of this enzyme may also play a role in ataxia-telangiectasia. Alternative splicing of this gene results in two transcript variants; however, the second variant has not yet been fully described.
Especificaciones

Especificaciones

IDH1
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
IDCD, IDH, IDH1, IDP, IDPC, NADP(+) specific ICDH, Oxalosuccinate decarboxylase, PICD
IDH1
IgG1
Protein G
Antibody
2A6A2
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human
O75874
3417
IDH1 Fusion Protein Ag19293
Primary
-20°C
IDH1
Videos
SDS
Documentos

Documentos

Product Certifications