Learn More
Novus Biologicals™ ICT Overexpression Lysate
Marca:  Novus Biologicals™ NBL1-11812
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18213315
  • 685.73€ / 0.10 miligramo
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
The ICT Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Especificaciones
Especificaciones
Western Blot
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Native
The lysate was created in HEK293T cells, using plasmid ID RC24448 and based on accession number NM_1545. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens immature colon carcinoma transcript 1 (ICT1), mRNA.
Cancer