missing translation for 'onlineSavingsMsg'
Learn More

ICOS (CD278) Armenian Hamster, PE-CF594, Clone: C398.4A, BD Horizon™

Armenian Hamster Monoclonal Antibody

Marca:  BD Biosciences 565889

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15824589

  • 183.00€ / cada uno


Fecha estimada de envío
07-01-2020
Añadir a la cesta

Descripción

Descripción

ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human, Mouse, Rat, Rhesus Ma samples. It is validated for Flow cytometry.
Especificaciones

Especificaciones

ICOS (CD278)
C398.4A
PE-CF594
Purified
Armenian Hamster
Armenian Hamster IgG
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed.
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
ICOS; AILIM; H4; Inducible T-cell costimulator; CVID1; Ly115
Mouse D10.G4.1 T-cell clone
25μg
Cell Biology
Primary
Human, Murine, Rat, Rhesus
Videos
Documentos

For Research Use Only.