Learn More
Gilac™ Ice Shovel
Product Code. 17938601
Change view
Click to view available options
Color:
White
Unit Size:
Each
Product Code. Color Unit Size
17938601 White Each
1 options
This item is not returnable. View return policy
Product Code. 17938601 Supplier Gilac™ Supplier No. G601021
  • ONLINE EXCLUSIVE PRICE
    190.10€
    124.58€ / paquete de 1
    Ahorro 65.52€
    34% Descuento
Fecha estimada de envoi: 01-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Ice Shovel is ideal for intensive use and has ergonomic handle.
  • Ideal for intensive use.
  • Ergonomic handle.
Specifications
Blade Material No Blade
Product Type One-piece Shovel
Blade Style No Blade
Color-coded No
For Use With (Equipment) Ice
Handle Material Polypropylene
Head Style Square
Length (English) 43.7 in.
Length (Metric) 1110 mm
Material Polypropylene
Color White