Learn More
ICAM-1/CD54 Antibody (1H4), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Bio-Techne NB110-58732FR
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18216085
  • 491.35€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock
Descripción
Descripción
ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface).
Especificaciones
Especificaciones
ICAM-1/CD54
Monoclonal
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
ICAM1
Raji cells and spleen cells fused with NS1 cells.
0.1 mL
Adaptive Immunity, Cancer, Cell Biology, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Stem Cell Markers
3383
Human
Purified
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
1H4
Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface)
BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
Mouse
Protein A purified
RUO
Primary
This antibody (clone 1H4) reacts with CD54 (ICAM-1), a 85-110 kDa type I transmembrane glycoprotein (receptor for rhinovirus) expressed on activated endothelial cells, T lymphocytes, B lymphocytes, monocytes, macrophages, granulocytes and dendritic cells; the expression of CD54 is upregulated by activation.
Store at 4C in the dark.
IgG2b