Learn More
Thermo Scientific™ HyperSep™ Pressure Manifold Collection Racks
Product Code. 13426926 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Collection Rack for 13 mm Tubes
Collection Rack for 16 mm Tubes
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
13426926 Collection Rack for 16 mm Tubes Each
15280349 Collection Rack for 13 mm Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13426926 Supplier Thermo Scientific™ Supplier No. 60104268
 Restricted Item
Fecha estimada de envoi: 22-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Use the collection rack that matches the size of solid collection tubes used with the Thermo Scientific™ HyperSep™ Positive Pressure Manifold.
Use the Thermo Scientific™ HyperSep™ Positive Pressure Manifold Collection Rack that matches the collection tube size being used. The HyperSep Positive Pressure Manifold prepares up to 48 samples by solid phase extraction (SPE) simultaneously. Extracts are collected into 13 or 16mm tubes.
The HyperSep Positive Pressure Manifold holds up to 48 SPE cartridges. Samples and solvent, added to the SPE cartridge, are extracted under positive pressure for an even flow at each position. Extracts are collected into 13 or 16mm tubes. This SPE manifold provides a quick, simple and robust way to prepare multiple samples for analysis by chromatography.
See related products if you need replacements for any of these HperSep Positive Pressure Manifold accessories.
TRUSTED_SUSTAINABILITY
Specifications
Description Collection Rack for 16 mm Tubes
For Use With (Application) Extract Collection
For Use With (Equipment) 16 mm Tubes, Positive Pressure Manifold
Product Line HyperSep
Type Collection Rack