Learn More
Safety First Aid HypaClens Eye Wash Dispenser
Beskrivelse
- The dispenser features clear instructions for using eye wash pods on front of the cabinet, enabling ordinary people to provide help in case of an emergency.
- The integrated mirror is ideal for self-administration of eye wash.
- The HypaClens 20 mL Eye Wash Dispenser is ideal in any workplace environment, including workplaces with higher number of employees.
- Refills can be purchased separately when required.
Tekniske data
Tekniske data
| Color | Clear |
| Flow Rate | 20 gpm (Shower), 8.0 gpm (Eye and Face Wash) |
| Material | Plastic |
| Mounting | Wall |
| Product Type | Portable Eyewash and Shower Fixtures |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?