Learn More
Hydroxy Tyrosol 3-Sulfate Sodium Salt, TRC
Product Code. 30012779
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012779 50 mg 50mg
30007778 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012779 Supplier Toronto Research Chemicals Supplier No. TRCH97701050MG
 Restricted Item
Fecha estimada de envoi: 10-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1391053-88-5
Molecular Formula C8H9NaO6S
Molecular Weight (g/mol) 256.21
Synonym 4-(2-Hydroxyethyl)-1,2-benzenediol 2-(Hydrogen Sulfate) Sodium;
IUPAC Name sodium 2-hydroxy-5-(2-hydroxyethyl)phenyl sulfate
SMILES OC1=CC=C(CCO)C=C1OS(=O)(O[Na])=O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C8H10O6S.Na/c9-4-3-6-1-2-7(10)8(5-6)14-15(11,12)13;/h1-2,5,9-10H,3-4H2,(H,11,12,13);/q;+1/p-1
Formula Weight 256
Chemical Name or Material Hydroxy Tyrosol 3-Sulfate Sodium Salt