missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNHIT2 Partial ORF (NP_055020.1, 274 a.a. - 361 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000741-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: GEHPPGPLGTRGAMHEVARILLGEGPTNQKGYTLAALGDLAQTLGRARKQAVAREERDHLYRARKKCQFLLAWTNENEAALTPLALDCEspecificaciones
NP_055020.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.42kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GEHPPGPLGTRGAMHEVARILLGEGPTNQKGYTLAALGDLAQTLGRARKQAVAREERDHLYRARKKCQFLLAWTNENEAALTPLALDC | |
RUO | |
ZNHIT2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
741 | |
ZNHIT2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C11orf5/FON/MGC120285/MGC120286 | |
ZNHIT2 | |
Recombinant | |
wheat germ expression system |