Learn More
Abnova™ Human ZNF331 Partial ORF (NP_061025.4, 362 a.a. - 459 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055422-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Zinc finger proteins have been shown to interact with nucleic acids and to have diverse functions. The zinc finger domain is a conserved amino acid sequence motif containing 2 specifically positioned cysteines and 2 histidines that are involved in coordinating zinc. Kruppel-related proteins form one family of zinc finger proteins. See ZFP93 (MIM 604749) for additional information on zinc finger proteins.[supplied by OMIM]
Sequence: KAFNCGYHLTQHERIHTGETPYKCKECGKAFIYGSSLVKHERIHTGVKPYGCTECGKSFSHGHQLTQHQKTHSGAKSYECKECGKACNHLNHLREHQREspecificaciones
NP_061025.4 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KAFNCGYHLTQHERIHTGETPYKCKECGKAFIYGSSLVKHERIHTGVKPYGCTECGKSFSHGHQLTQHQKTHSGAKSYECKECGKACNHLNHLREHQR | |
RUO | |
ZNF331 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
55422 | |
ZNF331 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686L0787/RITA/ZNF361/ZNF463 | |
ZNF331 | |
Recombinant | |
wheat germ expression system |