missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZC3H7A Full-length ORF (AAH12575.1, 1 a.a. - 167 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00029066-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16198952

  • 310.00€ / 10 microgramos


Fecha estimada de envío
12-02-2021
Añadir a la cesta

Descripción

Descripción

Sequence: MKENGIQDMEQFYELWLKSQKNEKSEDIASQSNKENGKQIHMPTDYAEVTVDFHCWMCGKNCNSEKQWQGHISSEKHKEKVFHTEDDQYCWQHRFPTGYFSICDRYMNGTCPEGNSCKFAHGNAELHEWEERRDALKMKLNKARKDHLIGPNDNDFGKYSFLFKDLN
Especificaciones

Especificaciones

AAH12575.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
46.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ10027|FLJ20318|HSPC055|ZC3H7|ZC3HDC7
ZC3H7A
Yes
wheat germ expression system
Liquid
29066
ZC3H7A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKENGIQDMEQFYELWLKSQKNEKSEDIASQSNKENGKQIHMPTDYAEVTVDFHCWMCGKNCNSEKQWQGHISSEKHKEKVFHTEDDQYCWQHRFPTGYFSICDRYMNGTCPEGNSCKFAHGNAELHEWEERRDALKMKLNKARKDHLIGPNDNDFGKYSFLFKDLN
RUO
ZC3H7A
Wheat Germ (in vitro)
GST
Videos
SDS
Documentos