missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZC3H7A Full-length ORF (AAH12575.1, 1 a.a. - 167 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00029066-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MKENGIQDMEQFYELWLKSQKNEKSEDIASQSNKENGKQIHMPTDYAEVTVDFHCWMCGKNCNSEKQWQGHISSEKHKEKVFHTEDDQYCWQHRFPTGYFSICDRYMNGTCPEGNSCKFAHGNAELHEWEERRDALKMKLNKARKDHLIGPNDNDFGKYSFLFKDLNEspecificaciones
AAH12575.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
46.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10027/FLJ20318/HSPC055/ZC3H7/ZC3HDC7 | |
ZC3H7A | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
29066 | |
ZC3H7A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKENGIQDMEQFYELWLKSQKNEKSEDIASQSNKENGKQIHMPTDYAEVTVDFHCWMCGKNCNSEKQWQGHISSEKHKEKVFHTEDDQYCWQHRFPTGYFSICDRYMNGTCPEGNSCKFAHGNAELHEWEERRDALKMKLNKARKDHLIGPNDNDFGKYSFLFKDLN | |
RUO | |
ZC3H7A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |