missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZBTB25 Partial ORF (NP_008908.2, 331 a.a. - 435 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00007597-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: DTEPVELNCNFSFSRKRKMSCTICGHKFPRKSQLLEHMYTHKGKSYRYNRCQRFGNALAQRFQPYCDSWSDVSLKSSRLSQEHLDLPCALESELTQENVDTILVEEspecificaciones
NP_008908.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.29kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DTEPVELNCNFSFSRKRKMSCTICGHKFPRKSQLLEHMYTHKGKSYRYNRCQRFGNALAQRFQPYCDSWSDVSLKSSRLSQEHLDLPCALESELTQENVDTILVE | |
RUO | |
ZBTB25 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7597 | |
ZBTB25 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KUP/ZNF46 | |
ZBTB25 | |
Recombinant | |
wheat germ expression system |