Learn More
Abnova™ Human YKT6 Full-length ORF (AAH07319, 1 a.a. - 198 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00010652-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene product is one of the SNARE recognition molecules implicated in vesicular transport between secretory compartments. It is a membrane associated, isoprenylated protein that functions at the endoplasmic reticulum-Golgi transport step. This protein is highly conserved from yeast to human and can functionally complement the loss of the yeast homolog in the yeast secretory pathway. [provided by RefSeq]
Sequence: MKLYSLSVLYKGEAKVVLLKAAYDVSSFSFFQRSSVQEFMTFTSQLIVERSSKGTRASVKEQDYLCHVYVRNDSLAGVVIADNEYPSRVAFTLLEKVLDEFSKQVDRIDWPVGSPATIHYPALDGHLSRYQNPREADPMTKVQAELDETKIILHNTMESLLERGEKLDDLVSKSEVLGTLSKAFYKTARKQNSCCAIMEspecificaciones
AAH07319 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
47.52kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
YKT6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10652 | |
YKT6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKLYSLSVLYKGEAKVVLLKAAYDVSSFSFFQRSSVQEFMTFTSQLIVERSSKGTRASVKEQDYLCHVYVRNDSLAGVVIADNEYPSRVAFTLLEKVLDEFSKQVDRIDWPVGSPATIHYPALDGHLSRYQNPREADPMTKVQAELDETKIILHNTMESLLERGEKLDDLVSKSEVLGTLSKAFYKTARKQNSCCAIM | |
RUO | |
YKT6 | |
Recombinant | |
wheat germ expression system |