Learn More
Abnova™ Human XRCC1 Partial ORF (NP_006288, 534 a.a. - 633 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00007515-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is involved in the efficient repair of DNA single-strand breaks formed by exposure to ionizing radiation and alkylating agents. This protein interacts with DNA ligase III, polymerase beta and poly (ADP-ribose) polymerase to participate in the base excision repair pathway. It may play a role in DNA processing during meiogenesis and recombination in germ cells. A rare microsatellite polymorphism in this gene is associated with cancer in patients of varying radiosensitivity. [provided by RefSeq]
Sequence: LPVPELPDFFQGKHFFLYGEFPGDERRKLIRYVTAFNGELEDYMSDRVQFVITAQEWDPSFEEALMDNPSLAFVRPRWIYSCNEKQKLLPHQLYGVVPQAEspecificaciones
NP_006288 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LPVPELPDFFQGKHFFLYGEFPGDERRKLIRYVTAFNGELEDYMSDRVQFVITAQEWDPSFEEALMDNPSLAFVRPRWIYSCNEKQKLLPHQLYGVVPQA | |
RUO | |
XRCC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7515 | |
XRCC1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RCC | |
XRCC1 | |
Recombinant | |
wheat germ expression system |