Learn More
Abnova™ Human WBSCR1 Partial ORF (NP_114381.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00007458-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes one of the translation initiation factors, which functions to stimulate the initiation of protein synthesis at the level of mRNA utilization. This gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene generates 2 transcript variants. [provided by RefSeq]
Sequence: MADFDTYDDRAYSSFGGGRGSRGSAGGHGSRSQKELPTEPPYTAYVGNLPFNTVQGDIDAIFKDLSIRSVRLVRDKDTDKFKGFCYVEFDEVDSLKEALTEspecificaciones
NP_114381.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MADFDTYDDRAYSSFGGGRGSRGSAGGHGSRSQKELPTEPPYTAYVGNLPFNTVQGDIDAIFKDLSIRSVRLVRDKDTDKFKGFCYVEFDEVDSLKEALT | |
RUO | |
EIF4H | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7458 | |
WBSCR1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0038/WBSCR1/WSCR1 | |
EIF4H | |
Recombinant | |
wheat germ expression system |