Learn More
Abcam Human WASF3 knockout HEK-293T cell line
Product Code. 32140735 Shop All Abcam Products
Change view
Click to view available options
Quantity:
2 x 1,000,000 Cells
Unit Size:
Pack of 2
Product Code. Quantity Unit Size
32140735 2 x 1,000,000 Cells Pack of 2
1 options
This item is not returnable. View return policy
Product Code. 32140735 Supplier Abcam Supplier No. ab2665682x1000000Cells_
 Restricted Item
Fecha estimada de envoi: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
WASF3 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: Insertion of the selection cassette in exon 7. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only kn...
WASF3 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: Insertion of the selection cassette in exon 7. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells
Regulatory Status RUO