missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human VPS37A Full-length ORF (AAH67754.1, 1 a.a. - 397 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00137492-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MSWLFPLTKSASSSAAGSPGGLTSLQQQKQRLIESLRNSHSSIAEIQKDVEYRLPFTINNLTININILLPPQFPQEKPVISVYPPIRHHLMDKQGVYVTSPLVNNFTMHSDLGKIIQSLLDEFWKNPPVLAPTSTAFPYLYSNPSGMSPYASQGFPFLPPYPPQEANRSITSLSVADTVSSSTTSHTTAKPAAPSFGVLSNLPLPIPTVDASIPTSQNGFGYKMPDVPDAFPELSELSVSQLTDMNEQEEVLLEQFLTLPQLKQIITDKDDLVKSIEELARKNLLLEPSLEAKRQTVLDKYELLTQMKSTFEKKMQRQHELSEICSASALQARLKVAAHEAEEESDNIAEDFLEGKMEIDDFLSSFMEKRTICHCRRAKEEKLQQAIAMHSQFHAPLEspecificaciones
AAH67754.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ32642/FLJ42616/HCRP1/PQBP2 | |
VPS37A | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
137492 | |
VPS37A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSWLFPLTKSASSSAAGSPGGLTSLQQQKQRLIESLRNSHSSIAEIQKDVEYRLPFTINNLTININILLPPQFPQEKPVISVYPPIRHHLMDKQGVYVTSPLVNNFTMHSDLGKIIQSLLDEFWKNPPVLAPTSTAFPYLYSNPSGMSPYASQGFPFLPPYPPQEANRSITSLSVADTVSSSTTSHTTAKPAAPSFGVLSNLPLPIPTVDASIPTSQNGFGYKMPDVPDAFPELSELSVSQLTDMNEQEEVLLEQFLTLPQLKQIITDKDDLVKSIEELARKNLLLEPSLEAKRQTVLDKYELLTQMKSTFEKKMQRQHELSEICSASALQARLKVAAHEAEEESDNIAEDFLEGKMEIDDFLSSFMEKRTICHCRRAKEEKLQQAIAMHSQFHAPL | |
RUO | |
VPS37A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |