missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human VPS11 Full-length ORF (AAH12051.1, 1 a.a. - 331 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055823-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MSEVQPDSPQGIYDTLLELRLQNWAHEKDPQVKEKLHAEAISLLKSGRFCDVFDKALVLCQMHDFQDGVLYLYEQGKLFQQIMHYHMQHEQYRQVISVCERHGEQDPSLWEQALSYFARKEEDCKEYVAAVLKHIENKNLMPPLLVVQTLAHNSTATLSVIRDYLVQKLQKQSQQIAQDELRVRRYREETTRIRQEIQELKASPKIFQKTKCSICNSALELPSVHFLCGHSFHQHCFESYSESDADCPTCLPENRKVMDMIRAQEQKRDLHDQFQHQLRCSNDSFSVIADYFGRGVFNKLTLLTDPPTARLTSSLEAGLQRDLLMHSRRGTEspecificaciones
AAH12051.1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
65kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
END1|PEP5|RNF108|hVPS11 | |
VPS11 | |
Yes |
wheat germ expression system | |
Liquid | |
55823 | |
VPS11 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSEVQPDSPQGIYDTLLELRLQNWAHEKDPQVKEKLHAEAISLLKSGRFCDVFDKALVLCQMHDFQDGVLYLYEQGKLFQQIMHYHMQHEQYRQVISVCERHGEQDPSLWEQALSYFARKEEDCKEYVAAVLKHIENKNLMPPLLVVQTLAHNSTATLSVIRDYLVQKLQKQSQQIAQDELRVRRYREETTRIRQEIQELKASPKIFQKTKCSICNSALELPSVHFLCGHSFHQHCFESYSESDADCPTCLPENRKVMDMIRAQEQKRDLHDQFQHQLRCSNDSFSVIADYFGRGVFNKLTLLTDPPTARLTSSLEAGLQRDLLMHSRRGT | |
RUO | |
VPS11 | |
Wheat Germ (in vitro) | |
GST |