Learn More
Abnova™ Human VAX2 Full-length ORF (NP_036608.1, 1 a.a. - 290 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00025806-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a homeobox protein and is almost exclusively expressed in the ventral portion of the retina during development. In mouse studies, this gene was found to be required for the correct formation of the optic fissure and other aspects of retinal development. [provided by RefSeq]
Sequence: MGDGGAERDRGPARRAESGGGGGRCGDRSGAGDLRADGGGHSPTEVAGTSASSPAGSRESGADSDGQPGPGEADHCRRILVRDAKGTIREIVLPKGLDLDRPKRTRTSFTAEQLYRLEMEFQRCQYVVGRERTELARQLNLSETQVKVWFQNRRTKQKKDQSRDLEKRASSSASEAFATSNILRLLEQGRLLSVPRAPSLLALTPSLPGLPASHRGTSLGDPRNSSPRLNPLSSASASPPLPPPLPAVCFSSAPLLDLPAGYELGSSAFEPYSWLERKVGSASSCKKANTEspecificaciones
NP_036608.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
57.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DRES93 | |
VAX2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
25806 | |
VAX2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGDGGAERDRGPARRAESGGGGGRCGDRSGAGDLRADGGGHSPTEVAGTSASSPAGSRESGADSDGQPGPGEADHCRRILVRDAKGTIREIVLPKGLDLDRPKRTRTSFTAEQLYRLEMEFQRCQYVVGRERTELARQLNLSETQVKVWFQNRRTKQKKDQSRDLEKRASSSASEAFATSNILRLLEQGRLLSVPRAPSLLALTPSLPGLPASHRGTSLGDPRNSSPRLNPLSSASASPPLPPPLPAVCFSSAPLLDLPAGYELGSSAFEPYSWLERKVGSASSCKKANT | |
RUO | |
VAX2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |