missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human UTP18 Full-length ORF (NP_057085.1, 1 a.a. - 521 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00051096-P01.2ug

Detalles adicionales : Peso : 0.00010kg

Código de producto. 16159602

  • 229.00€ / 2 microgramos
Fecha estimada de envío: 28-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Sequence: MRPDWKAGAGPGGPPQKPAPSSQRKPPARPSAAAAAIAVAAAEEERRLRQRNRLRLEEDKPAVERCLEELVFGDVENDEDALLRRLRGPRVQEHEDSGDSEVENEAKGNFPPQKKPVWVDEEDEDEEMVDMMNNRFRKDMMKNASESKLSKDNLKKRLKEEFQHAMGGVPAWAETTKRKTSSDDESEEDEDDLLQRTGNFISTSTSLPRGILKMKNCQHANAERPTVARISSVQFHPGAQIVMVAGLDNAVSLFQVDGKTNPKIQSIYLERFPIFKACFSANGEEVLATSTHSKVLYVYDMLAGKLIPVHQVRGLKEKIVRSFEVSPDGSFLLINGIAGYLHLLAMKTKELIGSMKINGRVAASTFSSDSKKVYASSGDGEVYVWDVNSRKCLNRFVDEGSLYGLSIATSRNGQYVACGSNCGVVNIYNQDSCLQETNPKPIKAIMNLVTGVTSLTFNPTTEILAIASEKMKEAVRLVHLPSCTVFSNFPVIKNKNISHVHTMDFSPRSGYFALGNEKARP
Especificaciones

Especificaciones

NP_057085.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
84.1kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CGI-48/WDR50
UTP18
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
51096
UTP18 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRPDWKAGAGPGGPPQKPAPSSQRKPPARPSAAAAAIAVAAAEEERRLRQRNRLRLEEDKPAVERCLEELVFGDVENDEDALLRRLRGPRVQEHEDSGDSEVENEAKGNFPPQKKPVWVDEEDEDEEMVDMMNNRFRKDMMKNASESKLSKDNLKKRLKEEFQHAMGGVPAWAETTKRKTSSDDESEEDEDDLLQRTGNFISTSTSLPRGILKMKNCQHANAERPTVARISSVQFHPGAQIVMVAGLDNAVSLFQVDGKTNPKIQSIYLERFPIFKACFSANGEEVLATSTHSKVLYVYDMLAGKLIPVHQVRGLKEKIVRSFEVSPDGSFLLINGIAGYLHLLAMKTKELIGSMKINGRVAASTFSSDSKKVYASSGDGEVYVWDVNSRKCLNRFVDEGSLYGLSIATSRNGQYVACGSNCGVVNIYNQDSCLQETNPKPIKAIMNLVTGVTSLTFNPTTEILAIASEKMKEAVRLVHLPSCTVFSNFPVIKNKNISHVHTMDFSPRSGYFALGNEKARP
RUO
UTP18
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales